4-Bromo-3,5-dihydroxybenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A18914-14 A18914-22
CAAAA18914-14EA 124.35 CAD
CAAAA18914-14 CAAAA18914-22 CAAAA18914-36
4-Bromo-3,5-dihydroxybenzoic acid 98+%
4-Bromo-3,5-dihydroxybenzoic acid

Purity: 98+% CAS No.: 16534-12-6 Molecular Formula: C7H5BrO4 Formula Weight: 233.02 Melting Point: 268-272° Beilstein Reference: 2368265

Formula: C₇H₅BrO₄
Melting Pt: 268…272 °C
Storage Temperature: Ambient
MDL Number: MFCD00002513
CAS Number: 16534-12-6
EINECS: 240-606-4
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR