6-Fluoro-m-anisaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18904-06
A18904-14
CAAAA18904-03EA
106.86
CAD
CAAAA18904-03
CAAAA18904-06
CAAAA18904-14
6-Fluoro-m-anisaldehyde 97%
6-Fluoro-m-anisaldehyde
Purity: 97%
CAS No.: 105728-90-3
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Flash Point: 107°(224°F)
Refractive Index: 1.5330
Boiling Point: 80°/0.9mm
Beilstein Reference: 4379088
MDL No.: MFCD00070795
Formula:
C₈H₇FO₂ Boiling Pt: 80 °C (0,9 mmHg) Density: 1,237 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070795 CAS Number: 105728-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...