4-Ethoxyphenyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethoxy-4-isothiocyanato-benzene, 4-Ethoxyphenylisothiocyanate

A18902-06 A18902-14
CAAAA18902-06EA 110.44 CAD
CAAAA18902-06 CAAAA18902-14
4-Ethoxyphenyl isothiocyanate 98%
4-Ethoxyphenyl isothiocyanate

Purity: 98% CAS No.: 3460-49-9 Molecular Formula: C9H9NOS Formula Weight: 179.24 Flash Point: 110°(230°F) Odor: Irritating Beilstein Reference: 637526 UN No.: 2923 MDL No.: MFCD00041096

Formula: C₉H₉NOS
MW: 179.24 g/mol
Boiling Pt: 146 °C (19 -rr)
Melting Pt: 57…61 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041096
CAS Number: 3460-49-9
EINECS: 222-410-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR