(±)-γ-Valerolactone 98+%
Supplier: Thermo Scientific Chemicals
A18872-14
A18872-22
A18872-36
CAAAAA18872-14EA
52.3
CAD
CAAAAA18872-14
CAAAA18872-22
CAAAA18872-36
(±)-γ-Valerolactone 98+%
(±)-γ-Valerolactone
Purity: 98+%
CAS No.: 108-29-2
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 81°(178°F)
Refractive Index: 1.4330
Odor: Pleasant
Melting Point: -31°
Boiling Point: 207-208°
Beilstein Reference: 80420
MDL No.: MFCD00005400
Formula:
C₅H₈O₂ Boiling Pt: 207…208 °C Melting Pt: –31 °C Density: 1.052 Flash Pt: 81 °C(178 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005400 CAS Number: 108-29-2 EINECS: 203-569-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...