1,8-Naphthalimide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Naphthalene-1,8-dicarboximide

A18856-09 A18856-18
CAAAAA18856-30EA 878.46 CAD
CAAAAA18856-30 CAAAA18856-09 CAAAA18856-18
1,8-Naphthalimide 98%
1,8-Naphthalimide

Purity: 98% CAS No.: 81-83-4 Molecular Formula: C12H7NO2 Formula Weight: 197.20 Melting Point: 299-303° Beilstein Reference: 153150

Formula: C₁₂H₇NO₂
MW: 197.19 g/mol
Boiling Pt: 475
Melting Pt: 299…303 °C
Storage Temperature: Ambient
MDL Number: MFCD00006920
CAS Number: 81-83-4
EINECS: 201-379-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR