cis-2-Nonen-1-ol 95%
Supplier: Thermo Scientific Chemicals
A18842-06
A18842-14
CAAAA18842-06EA
96.85
CAD
CAAAA18842-06
CAAAA18842-14
cis-2-Nonen-1-ol 95%
cis-2-Nonen-1-ol
Purity: 95%
CAS No.: 41453-56-9
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 101°(214°F)
Refractive Index: 1.4500
Boiling Point: 106°/20mm
MDL No.: MFCD00063210
Formula:
C₉H₁₈O Boiling Pt: 106 °C/20 mm Density: 0.845 Flash Pt: 101 °C(214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063210 CAS Number: 41453-56-9 EINECS: 255-376-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...