1-Methylcyclopentene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18816-06
A18816-14
A18816-22
CAAAA18816-06EA
111.55
CAD
CAAAA18816-06
CAAAA18816-14
CAAAA18816-22
1-Methylcyclopentene 98%
1-Methylcyclopentene
Purity: 98%
CAS No.: 693-89-0
Molecular Formula: C6H10
Formula Weight: 82.15
Flash Point: -17°(1°F)
Refractive Index: 1.4310
Melting Point: -142°
Boiling Point: 73-75°
Beilstein Reference: 1900640
UN No.: 3295
MDL No.: MFCD00001397
Formula:
C₆H₁₀ Boiling Pt: 73…75 °C Melting Pt: –142 °C Density: 0.779 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001397 CAS Number: 693-89-0 EINECS: 211-762-0 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...