Decyl acetate 98%
Supplier: Thermo Scientific Chemicals
Warning
|
A18814-0B
A18814-30
CAAAAA18814-36EA
139.13
CAD
CAAAAA18814-36
CAAAA18814-0B
CAAAA18814-30
Decyl acetate 98%
Decyl acetate
Purity: 98%
CAS No.: 112-17-4
Molecular Formula: CH3CO2(CH2)9CH3
Formula Weight: 200.32
Flash Point: 125°(257°F)
Refractive Index: 1.4270
Boiling Point: 244°
Beilstein Reference: 1762123
MDL No.: MFCD00026538
Formula:
C₁₂H₂₄O₂ Boiling Pt: 244 °C Density: 0.867 Flash Pt: 125 °C(257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026538 CAS Number: 112-17-4 EINECS: 203-942-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...