(-)-Cinchonidine 99% (total base) may cont. up to 5% quinine
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18796-14
A18796-22
CAAAA18796-14EA
105.23
CAD
CAAAA18796-14
CAAAA18796-22
(-)-Cinchonidine 99% (total base) may cont. up to 5% quinine
(-)-Cinchonidine
Purity: 99%
CAS No.: 485-71-2
Molecular Formula: C19H22N2O
Formula Weight: 294.39
Melting Point: 201-206°
Beilstein Reference: 89690
Merck Reference: 14,2286
Optical Rotation: -109° (c=0.5 in ethanol)
MDL No.: MFCD00006783
Formula:
C₁₉H₂₂N₂O Boiling Pt: 465 °C (1013 hPa) Melting Pt: 201…206 °C Density: 1.22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006783 CAS Number: 485-71-2 EINECS: 207-622-3 Merck Index: 14,2286 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...