3,4-Dinitrobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A18774-06 A18774-14 A18774-22
CAAAAA18774-36EA 2083.69 CAD
CAAAAA18774-36 CAAAA18774-06 CAAAA18774-14 CAAAA18774-22
3,4-Dinitrobenzoic acid 99%
3,4-Dinitrobenzoic acid

Purity: 99% CAS No.: 528-45-0 Molecular Formula: C7H4N2O6 Formula Weight: 212.12 Melting Point: 163-167° Beilstein Reference: 2055333 Merck Reference: 14,3275 MDL No.: MFCD00007255

Formula: C₇H₄N₂O₆
Melting Pt: 163…167 °C
Density: 1.674 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007255
CAS Number: 528-45-0
EINECS: 208-433-9
Merck Index: 14,3275

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR