2-Indolinone 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone

A18764-22 A18764-06 A18764-14
CAAAAA18764-22EA 726.8 CAD
CAAAAA18764-22 CAAAA18764-06 CAAAA18764-14
2-Indolinone 97+%
2-Indolinone

Purity: 97+% CAS No.: 59-48-3 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 124-128° Boiling Point: 194-196°/17mm MDL No.: MFCD00005711

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 194…196 °C/17 mm
Melting Pt: 124…128 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR