2-Amino-4-ethylpyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

A18754-09 A18754-88
CAAAA18754-09EA 350.76 CAD
CAAAA18754-09 CAAAA18754-88
2-Amino-4-ethylpyridine 97%
2-Amino-4-ethylpyridine

Purity: 97% CAS No.: 33252-32-3 Molecular Formula: C7H10N2 Formula Weight: 122.17 Melting Point: 65-69° UN No.: 2811 MDL No.: MFCD00129025

Formula: C₇H₁₀N₂
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00129025
CAS Number: 33252-32-3
EINECS: 251-430-2
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR