3',4'-Dichloropropiophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3,4-Dichlorophenyl)-1-propanone, 1-(3,4-Dichlorophenyl)propan-1-one

A18740-06 A18740-14
CAAAA18740-06EA 85.95 CAD
CAAAA18740-06 CAAAA18740-14
3',4'-Dichloropropiophenone 98%
3',4'-Dichloropropiophenone

Purity: 98% CAS No.: 6582-42-9 Molecular Formula: C9H8Cl2O Formula Weight: 203.07 Flash Point: 110°(230°F) Melting Point: 44-46° Boiling Point: 136-140°/18mm MDL No.: MFCD00000554

Formula: C₉H₈Cl₂O
MW: 203.07 g/mol
Boiling Pt: 136…140 °C/18 mm
Melting Pt: 44…46 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000554
CAS Number: 6582-42-9
EINECS: 229-511-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR