2,2,6,6-Tetramethylpiperidine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18712-22
A18712-06
A18712-14
CAAAAA18712-22EA
682.56
CAD
CAAAAA18712-22
CAAAA18712-06
CAAAA18712-14
2,2,6,6-Tetramethylpiperidine 98+%
2,2,6,6-Tetramethylpiperidine
Purity: 98+%
CAS No.: 768-66-1
Molecular Formula: C9H19N
Formula Weight: 141.26
Flash Point: 34°(93°F)
Refractive Index: 1.4440
Odor: Amine-like
Melting Point: -59°
Boiling Point: 152-154°
Beilstein Reference: 103296
UN No.: 2733
MDL No.: MFCD00005985
Formula:
C₉H₁₉N MW: 141.26 g/mol Boiling Pt: 152…154 °C Melting Pt: –59 °C Density: 0.832 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005985 CAS Number: 768-66-1 EINECS: 212-199-3 UN: 1992 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...