1-Bromo-2-naphthol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromonaphthalen-2-ol, 1-Bromo-2-hydroxynaphthalene

A18692-14 A18692-22
CAAAA18692-14EA 90.85 CAD
CAAAA18692-14 CAAAA18692-22
1-Bromo-2-naphthol 97%
1-Bromo-2-naphthol

Purity: 97% CAS No.: 573-97-7 Molecular Formula: C10H7BrO Formula Weight: 223.08 Melting Point: 76-81° Beilstein Reference: 2044001

Formula: C₁₀H₇BrO
MW: 223.07 g/mol
Boiling Pt: 314
Melting Pt: 76…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00003869
CAS Number: 573-97-7
EINECS: 209-363-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR