4'-Bromo-2'-chloroacetanilide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A18652-14 A18652-22
CAAAA18652-14EA 178.54 CAD
CAAAA18652-14 CAAAA18652-22
4'-Bromo-2'-chloroacetanilide 98+%
4'-Bromo-2'-chloroacetanilide
Formula: H₃CC(O)NH(C₆H₃BrCl)
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00040852
CAS Number: 3460-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR