Isovaleric acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18642-AK
A18642-AU
CAAAA18642-AKEA
48.98
CAD
CAAAA18642-AK
CAAAA18642-AU
Isovaleric acid 98%
Isovaleric acid
Purity: 98%
CAS No.: 503-74-2
Molecular Formula: (CH3)2CHCH2CO2H
Formula Weight: 102.13
Flash Point: 70°(158°F)
Refractive Index: 1.4030
Odor: Rancid
pH: 3
Melting Point: -33°
Boiling Point: 175-177°
Beilstein Reference: 1098522
Merck Reference: 14,5231
UN No.: 3265
MDL No.: MFCD00002726
Formula:
C₅H₁₀O₂ Boiling Pt: 175…177 °C Melting Pt: –33 °C Density: 0.937 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002726 CAS Number: 503-74-2 EINECS: 207-975-3 UN: 3265 ADR: 8,III Merck Index: 14,5231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...