Indole-5-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18627-03
A18627-06
A18627-14
CAAAA18627-03EA
74.42
CAD
CAAAA18627-03
CAAAA18627-06
CAAAA18627-14
Indole-5-carboxylic acid 98%
Indole-5-carboxylic acid
Purity: 98%
CAS No.: 1670-81-1
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Melting Point: 208-212°
Beilstein Reference: 124391
Formula:
C₉H₇NO₂ Melting Pt: 208…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005678 CAS Number: 1670-81-1 EINECS: 216-799-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...