Methyl chlorodifluoroacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18623-09
A18623-30
CAAAA18623-09EA
51.82
CAD
CAAAA18623-09
CAAAA18623-18
CAAAA18623-30
Methyl chlorodifluoroacetate 98%
Methyl chlorodifluoroacetate
Purity: 98%
CAS No.: 1514-87-0
Molecular Formula: ClCF2CO2CH3
Formula Weight: 144.50
Flash Point: 19°(66°F)
Refractive Index: 1.3500
Boiling Point: 79-81°
Beilstein Reference: 1756072
UN No.: 2924
MDL No.: MFCD00000775
Formula:
C₃H₃ClF₂O₂ Boiling Pt: 79…81 °C Density: 1.371 Flash Pt: 19 °C (66 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000775 CAS Number: 1514-87-0 EINECS: 216-154-9 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...