Isophytol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18583-09
A18583-18
CAAAA18583-09EA
66.52
CAD
CAAAA18583-09
CAAAA18583-18
Isophytol 95%
Isophytol
Purity: 95%
CAS No.: 505-32-8
Molecular Formula: C20H40O
Formula Weight: 296.54
Refractive Index: 1.4571
Boiling Point: 125-128°/0.06mm
Merck Reference: 14,5198
MDL No.: MFCD00048380
Formula:
C₂₀H₄₀O Boiling Pt: 125…128 °C/0.06 mm Density: 0.8519 Storage Temperature: Ambient |
MDL Number:
MFCD00048380 CAS Number: 505-32-8 EINECS: 208-008-8 Merck Index: 14,5198 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...