4,4'-Dimethylbenzil 98%

Supplier: Thermo Scientific Chemicals

A18576-03 A18576-06 A18576-14
CAAAA18576-03EA 49.61 CAD
CAAAA18576-03 CAAAA18576-06 CAAAA18576-14
4,4'-Dimethylbenzil 98%
4,4'-Dimethylbenzil

Purity: 98% CAS No.: 3457-48-5 Molecular Formula: C16H14O2 Formula Weight: 238.29 Melting Point: 102-104° MDL No.: MFCD00008554

Formula: C₁₆H₁₄O₂
Boiling Pt: 209 - 211 °C (7 -rr)
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00008554
CAS Number: 3457-48-5
EINECS: 222-388-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR