1,2-Cyclohexanediol cis- and trans-mixture 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Cyclohexane-1,2-diol

A18572-09 A18572-18
CAAAA18572-09EA 90.69 CAD
CAAAA18572-09 CAAAA18572-18
1,2-Cyclohexanediol cis- and trans-mixture 98%
1,2-Cyclohexanediol

Purity: 98% CAS No.: 931-17-9 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 110°(230°F) Melting Point: 72-76° Boiling Point: 118-120°/10mm MDL No.: MFCD00003861

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 118…120 °C/10 mm
Melting Pt: 72…76 °C
Density: 0.948 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003861
CAS Number: 931-17-9
EINECS: 213-229-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR