Sodium alginate, very low viscosity

Supplier: Thermo Scientific Chemicals

A18565-0E A18565-22 A18565-36
CAAAA18565-0EEA 900.6 CAD
CAAAA18565-0E CAAAA18565-22 CAAAA18565-36
Sodium alginate, very low viscosity
Sodium alginate

CAS No.: 9005-38-3 Merck Reference: 14,241 MDL No.: MFCD00081310

Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Merck Index: 14,241

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR