2-Amino-6-bromobenzothiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

A18554-03 A18554-06 A18554-14
CAAAA18554-03EA 87.06 CAD
CAAAA18554-03 CAAAA18554-06 CAAAA18554-14
2-Amino-6-bromobenzothiazole 98%
2-Amino-6-bromobenzothiazole

Purity: 98% CAS No.: 15864-32-1 Molecular Formula: C7H5BrN2S Formula Weight: 229.10 Odor: Odorless

Formula: C₇H₅BrN₂S
MW: 229.1
Storage Temperature: Ambient
MDL Number: MFCD00152229
CAS Number: 15864-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR