β-Naphthoflavone 98+%

Supplier: Thermo Scientific Chemicals

A18543-03 A18543-06
CAAAA18543-03EA 70.31 CAD
CAAAA18543-03 CAAAA18543-06
β-Naphthoflavone 98+%
β-Naphthoflavone

Inhibitor of aryl hydrocarbon hydroxylase


Purity: 98+% CAS No.: 6051-87-2 Molecular Formula: C19H12O2 Formula Weight: 272.31 Odor: Odorless Melting Point: 162-166° Beilstein Reference: 18991 MDL No.: MFCD00004986

Formula: C₁₉H₁₂O₂
Boiling Pt: 461 °C (1013 hPa)
Melting Pt: 162…166 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004986
CAS Number: 6051-87-2
EINECS: 227-958-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR