2'-Hydroxy-6'-methoxyacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Hydroxy-6-methoxyphenyl)ethanone

A18539-03 A18539-06 A18539-14
CAAAA18539-03EA 54.83 CAD
CAAAA18539-03 CAAAA18539-06 CAAAA18539-14
2'-Hydroxy-6'-methoxyacetophenone 97%
2'-Hydroxy-6'-methoxyacetophenone

Purity: 97% CAS No.: 703-23-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 56-59° Boiling Point: 141°/16mm MDL No.: MFCD00008732

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 141 °C (16 mmHg)
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00008732
CAS Number: 703-23-1
EINECS: 211-872-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR