7-Hydroxyflavone 98%
Supplier: Thermo Scientific Chemicals
A18538-03
A18538-06
CAAAA18538-03EA
85.16
CAD
CAAAA18538-03
CAAAA18538-06
7-Hydroxyflavone 98%
7-Hydroxyflavone
Purity: 98%
CAS No.: 6665-86-7
Molecular Formula: C15H10O3
Formula Weight: 238.25
Melting Point: 245-247°
Beilstein Reference: 194089
Formula:
C₁₅H₁₀O₃ Boiling Pt: 395 °C (1013 hPa) Melting Pt: 245…247 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006835 CAS Number: 6665-86-7 EINECS: 229-705-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...