1-Acetyl-2-naphthol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Acetyl-2-hydroxynaphthalene, 2-Hydroxy-1-acetonaphthone, 1-(2-Hydroxynaphthalen-1-yl)ethanone, 2'-Hydroxy-1'-acetonaphthone, 1-(2-Hydroxy-1-naphthyl)ethan-1-one

A18537-06 A18537-14
CAAAAA18537-06EA 39.34 CAD
CAAAAA18537-06 CAAAA18537-14 CAAAA18537-22
1-Acetyl-2-naphthol 99%
1-Acetyl-2-naphthol

Purity: 99% CAS No.: 574-19-6 Molecular Formula: C12H10O2 Formula Weight: 186.21 Melting Point: 63-67° Boiling Point: 130-132°/2mm MDL No.: MFCD00012131

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 178
Melting Pt: 64
Storage Temperature: Ambient
MDL Number: MFCD00012131
CAS Number: 574-19-6
EINECS: 209-369-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR