4-Nitrobenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Nitrobenzene-1-sulfonyl chloride, 4-Nitrobenzene-1-sulphonyl chloride

A18512-36 A18512-14 A18512-22
CAAAAA18512-36EA 1090.2 CAD
CAAAAA18512-36 CAAAA18512-14 CAAAA18512-22
4-Nitrobenzenesulfonyl chloride 97%
4-Nitrobenzenesulphonyl chloride

Purity: 97% CAS No.: 98-74-8 Molecular Formula: C6H4ClNO4S Formula Weight: 221.62 Melting Point: 76-80° Boiling Point: 143-144°/1.5mm UN No.: 3261 MDL No.: MFCD00007445

Formula: C₆H₄ClNO₄S
MW: 221.62 g/mol
Boiling Pt: 143…144 °C/1.5 mm
Melting Pt: 76…80 °C
Density: 1.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007445
CAS Number: 98-74-8
EINECS: 202-697-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR