2,4-Di-tert-butylphenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18509-0B
A18509-30
CAAAA18509-0BEA
127.35
CAD
CAAAA18509-0B
CAAAA18509-30
2,4-Di-tert-butylphenol 97%
2,4-Di-tert-butylphenol
Purity: 97%
CAS No.: 96-76-4
Molecular Formula: C14H22O
Formula Weight: 206.33
Flash Point: 129°(264°F)
Melting Point: 53-57°
Boiling Point: 265°
Beilstein Reference: 1910383
UN No.: 3077
MDL No.: MFCD00008828
Formula:
C₁₄H₂₂O Boiling Pt: 265 °C Melting Pt: 53…57 °C Density: 0.887 Flash Pt: 129 °C (264 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008828 CAS Number: 96-76-4 EINECS: 202-532-0 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...