Benzyl nicotinate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18505-18
A18505-30
CAAAA18505-18EA
96.06
CAD
CAAAA18505-18
CAAAA18505-30
Benzyl nicotinate 99%
Benzyl nicotinate
Purity: 99%
CAS No.: 94-44-0
Molecular Formula: C13H11NO2
Formula Weight: 213.24
Flash Point: 170°(338°F)
Refractive Index: 1.5700
Melting Point: 21-23°
Boiling Point: 179-180°/5mm
Beilstein Reference: 159169
Merck Reference: 14,6526
MDL No.: MFCD00023584
Formula:
C₁₃H₁₁NO₂ Boiling Pt: 179…180 °C/5 mm Melting Pt: 21…23 °C Density: 1.165 Flash Pt: 170 °C(338 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00023584 CAS Number: 94-44-0 EINECS: 202-332-3 Merck Index: 14,6526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...