2-Fluoro-4-(trifluoromethyl)benzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18500-14
A18500-03
A18500-06
CAAAAA18500-14EA
704.68
CAD
CAAAAA18500-14
CAAAA18500-03
CAAAA18500-06
2-Fluoro-4-(trifluoromethyl)benzaldehyde 97%
2-Fluoro-4-(trifluoromethyl)benzaldehyde
Purity: 97%
CAS No.: 89763-93-9
Molecular Formula: C8H4F4O
Formula Weight: 192.11
Flash Point: 110° (230°F)
Refractive Index: 1.4500
Boiling Point: 118-119°
Beilstein Reference: 7703632
MDL No.: MFCD00061310
Formula:
C₈H₄F₄O MW: 192.11 g/mol Boiling Pt: 118…119 °C Density: 1.410 Flash Pt: 110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00061310 CAS Number: 89763-93-9 UN: 1987 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...