2-Naphthyl acetate 99%
Supplier: Thermo Scientific Chemicals
A18487-30
A18487-09
A18487-18
CAAAAA18487-30EA
562.48
CAD
CAAAAA18487-30
CAAAA18487-09
CAAAA18487-18
2-Naphthyl acetate 99%
2-Naphthyl acetate
Purity: 99%
CAS No.: 1523-11-1
Molecular Formula: C12H10O2
Formula Weight: 186.21
Storage Temperature: 2°C to 8°C
Melting Point: 68-70°
Beilstein Reference: 1868291
MDL No.: MFCD00004060
Formula:
C₁₂H₁₀O₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 68…70 °C Density: 1.1108 g/cm³ (70 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004060 CAS Number: 1523-11-1 EINECS: 216-194-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...