Bromophenol blue

Supplier: Thermo Scientific Chemicals

A18469-09 A18469-18
CAAAA18469-09EA 71.89 CAD
CAAAA18469-09 CAAAA18469-18
Bromophenol blue
Bromophenol blue

CAS No.: 115-39-9 Molecular Formula: C19H10Br4O5S Formula Weight: 669.98 Odor: Odorless Melting Point: ca 270° dec. Beilstein Reference: 61698 Merck Reference: 14,1444 MDL No.: MFCD00005875

Formula: C₁₉H₁₀Br₄O₅S
Melting Pt: ∼ 270 °C (dec.)
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 14,1444

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR