1,5-Dibromopentane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18465-0E
A18465-22
A18465-36
CAAAAA18465-0EEA
1009.62
CAD
CAAAAA18465-0E
CAAAA18465-22
CAAAA18465-36
1,5-Dibromopentane 98%
1,5-Dibromopentane
Purity: 98%
CAS No.: 111-24-0
Molecular Formula: Br(CH2)5Br
Formula Weight: 229.95
Flash Point: 79°(174°F)
Refractive Index: 1.5122
Melting Point: -34°
Boiling Point: 224°
Beilstein Reference: 1209245
MDL No.: MFCD00000268
Formula:
BrH₂C(CH₂)₃CH₂Br Boiling Pt: 224 °C Melting Pt: –34 °C Density: 1.688 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000268 CAS Number: 111-24-0 EINECS: 203-849-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...