2-sec-Butylphenol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18461-0B
A18461-30
CAAAA18461-0BEA
113.29
CAD
CAAAA18461-0B
CAAAA18461-30
2-sec-Butylphenol 98%
2-sec-Butylphenol
Purity: 98%
CAS No.: 89-72-5
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 112°(233°F)
Refractive Index: 1.5220
Odor: Phenol-like
Melting Point: 10-12°
Boiling Point: 226-228°
Beilstein Reference: 1210026
UN No.: 3145
MDL No.: MFCD00002225
Formula:
C₁₀H₁₄O Boiling Pt: 226…228 °C Melting Pt: 10…12 °C Density: 0.982 Flash Pt: 112 °C (233 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002225 CAS Number: 89-72-5 EINECS: 201-933-8 UN: 3145 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...