Di-isopropanolamine sum of isomers 98+% (sum of isomers)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1'-Iminodipropan-2-ol, Bis(2-hydroxypropyl)amine, 1,1-Iminodi-2-propanol, 1-[(2-Hydroxypropyl)amino]propan-2-ol

A18451-0B A18451-30
CAAAA18451-0BEA 143.15 CAD
CAAAA18451-0B CAAAA18451-30
Di-isopropanolamine sum of isomers 98+% (sum of isomers)
Di-isopropanolamine

Purity: 98+% CAS No.: 110-97-4 Molecular Formula: C6H15NO2 Formula Weight: 133.19 Flash Point: 130°(266°F) Odor: Amine-like Boiling Point: 249-250° Beilstein Reference: 605363 Merck Reference: 14,3195 MDL No.: MFCD00004531

Formula: C₆H₁₅NO₂
Boiling Pt: 249…250 °C
Melting Pt: 38…45 °C
Density: 1.004
Flash Pt: 130 °C(266 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004531
CAS Number: 110-97-4
EINECS: 203-820-9
Merck Index: 14,3195

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR