Dibenzyl ether 98+%

Supplier: Thermo Scientific Chemicals
Warning

A18447-30 A18447-0E A18447-36
CAAAAA18447-30EA 46.29 CAD
CAAAAA18447-30 CAAAA18447-0E CAAAA18447-36
Dibenzyl ether 98+%
Dibenzyl ether

Purity: 98+% CAS No.: 103-50-4 Molecular Formula: C6H5CH2OCH2C6H5 Formula Weight: 198.27 Flash Point: 135°(275°F) Refractive Index: 1.5620 Melting Point: 3-4° Boiling Point: 298° Beilstein Reference: 1911156 Merck Reference: 14,1132 UN No.: 3082 MDL No.: MFCD00004780

Formula: C₆H₅CH₂OCH₂C₆H₅
Boiling Pt: 298 °C
Melting Pt: 3…4 °C
Density: 1.040
Flash Pt: 135 °C (275 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004780
CAS Number: 103-50-4
EINECS: 203-118-2
UN: 3082
ADR: 9,III
Merck Index: 14,1132
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cb077d69cfe8c3315f62058a54ca8460d752735&vid=08fffe90d8cf2c6ddb596093c7e9631247e2a70b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR