2,3-Dichloro-1-propanol 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18444-06
A18444-14
CAAAA18444-06EA
76
CAD
CAAAA18444-06
CAAAA18444-14
2,3-Dichloro-1-propanol 97+%
2,3-Dichloro-1-propanol
Purity: 97+%
CAS No.: 616-23-9
Molecular Formula: ClCH2CHClCH2OH
Formula Weight: 128.99
Flash Point: 93°(199°F)
Refractive Index: 1.4850
Boiling Point: 182°
Beilstein Reference: 1732060
UN No.: 2810
MDL No.: MFCD00040446
Formula:
C₃H₆Cl₂O Boiling Pt: 182 °C Melting Pt: –44 °C Density: 1.361 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040446 CAS Number: 616-23-9 EINECS: 210-470-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...