5-Iodo-m-xylene 97%

Supplier: Thermo Scientific Chemicals
Warning

A18413-09 A18413-18
CAAAA18413-09EA 82.63 CAD
CAAAA18413-09 CAAAA18413-18
5-Iodo-m-xylene 97%
5-Iodo-m-xylene

Purity: 97% CAS No.: 22445-41-6 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 110°(230°F) Refractive Index: 1.5950 Boiling Point: 229° Beilstein Reference: 2039353 MDL No.: MFCD00060659

Formula: C₈H₉I
Boiling Pt: 229 °C
Density: 1.610
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060659
CAS Number: 22445-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR