Dibenzosuberone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Dibenzo[b,f]cycloheptanone, 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one, 5-Dibenzosuberone, Dibenzo[b,f]cycloheptan-1-one
A18405-14
A18405-22
CAAAAA18405-14EA
60.67
CAD
CAAAAA18405-14
CAAAAA18405-22
CAAAA18405-30
Dibenzosuberone 97%
Dibenzosuberone
Purity: 97%
CAS No.: 1210-35-1
Molecular Formula: C15H12O
Formula Weight: 208.26
Flash Point: 200°(392°F)
Refractive Index: 1.6340
Melting Point: 32-34°
Boiling Point: 147-149°/0.3mm
Beilstein Reference: 1454355
MDL No.: MFCD00003587
Formula:
C₁₅H₁₂O MW: 208.26 g/mol Boiling Pt: 147…149 °C/0.3 mm Melting Pt: 32…34 °C Density: 1.156 Flash Pt: 200 °C(392 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003587 CAS Number: 1210-35-1 EINECS: 214-912-3 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...