(±)-2-Heptanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(±)-Methyl pentyl carbinol, (±)-2-Hydroxy heptane, Heptan-2-ol, (±)-Pentyl methyl carbinol
A18391-18
A18391-30
CAAAA18391-18EA
56.09
CAD
CAAAA18391-18
CAAAA18391-30
(±)-2-Heptanol 98%
(±)-2-Heptanol
Purity: 98%
CAS No.: 543-49-7
Molecular Formula: C7H16O
Formula Weight: 116.20
Flash Point: 59°(138°F)
Refractive Index: 1.4210
Boiling Point: 160-162°
Beilstein Reference: 1719088
Merck Reference: 14,4662
UN No.: 1987
MDL No.: MFCD00004587
Formula:
C₇H₁₆O MW: 116.2 g/mol Boiling Pt: 160 Density: 0.819 Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004587 CAS Number: 543-49-7 EINECS: 208-844-3 UN: 1987 ADR: 3 III Merck Index: 14,4662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...