2-Bromo-2-nitropropane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18385-06
A18385-14
CAAAA18385-06EA
48.98
CAD
CAAAA18385-06
CAAAA18385-14
2-Bromo-2-nitropropane 98%
2-Bromo-2-nitropropane
Purity: 98%
CAS No.: 5447-97-2
Molecular Formula: C3H6BrNO2
Formula Weight: 168.00
Flash Point: 61°(141°F)
Refractive Index: 1.4640
Boiling Point: 45-46°/16mm
Beilstein Reference: 1746626
MDL No.: MFCD00007389
Formula:
H₃CBrC(NO₂)CH₃ Boiling Pt: 45…46 °C/16 mm Melting Pt: 33…35 °C Density: 1,584 Flash Pt: 61 °C(141 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007389 CAS Number: 5447-97-2 EINECS: 226-668-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...