1-Fluoronaphthalene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18378-06
A18378-14
A18378-22
CAAAA18378-06EA
56.25
CAD
CAAAA18378-06
CAAAA18378-14
CAAAA18378-22
1-Fluoronaphthalene 98%
1-Fluoronaphthalene
Purity: 98%
CAS No.: 321-38-0
Molecular Formula: C10H7F
Formula Weight: 146.17
Flash Point: 65°(149°F)
Refractive Index: 1.5930
Melting Point: -10°
Boiling Point: 215-217°
Beilstein Reference: 1906413
MDL No.: MFCD00003873
Formula:
C₁₀H₇F Boiling Pt: 215…217 °C Melting Pt: –10 °C Density: 1.331 Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003873 CAS Number: 321-38-0 EINECS: 206-287-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...