1-Methoxy-2,2,2-trifluoroethanol tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trifluoroacetaldehyde methyl hemiacetal

A18369-06 A18369-14 A18369-22
CAAAA18369-06EA 61.62 CAD
CAAAA18369-06 CAAAA18369-14 CAAAA18369-22
1-Methoxy-2,2,2-trifluoroethanol tech. 90%, Technical Grade
1-Methoxy-2,2,2-trifluoroethanol

Purity: 90% CAS No.: 431-46-9 Molecular Formula: C3H5F3O2 Formula Weight: 130.07 Flash Point: 42°(107°F) Refractive Index: 1.3330 Boiling Point: 96° Beilstein Reference: 1737758 UN No.: 3271 MDL No.: MFCD00013572

Formula: C₃H₅F₃O₂
MW: 130.07 g/mol
Boiling Pt: 96 °C
Density: 1.360
Flash Pt: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013572
CAS Number: 431-46-9
EINECS: 207-072-4
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR