3,5-Dibromo-4-hydroxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A18361-14 A18361-22
CAAAA18361-14EA 72.84 CAD
CAAAA18361-14 CAAAA18361-22
3,5-Dibromo-4-hydroxybenzoic acid 98%
3,5-Dibromo-4-hydroxybenzoic acid

Purity: 98% CAS No.: 3337-62-0 Molecular Formula: C7H4Br2O3 Formula Weight: 295.93 Melting Point: 269-273° Beilstein Reference: 2416147

Formula: C₇H₄Br₂O₃
MW: 295.91 g/mol
Boiling Pt: 365
Melting Pt: 269…273 °C
Storage Temperature: Ambient
MDL Number: MFCD00002548
CAS Number: 3337-62-0
EINECS: 222-075-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR