2-Hydroxybenzimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

A18324-22 A18324-09 A18324-18 A18324-30
CAAAAA18324-22EA 383.94 CAD
CAAAAA18324-22 CAAAA18324-09 CAAAA18324-18 CAAAA18324-30
2-Hydroxybenzimidazole 98%
2-Hydroxybenzimidazole

Purity: 98% CAS No.: 615-16-7 Molecular Formula: C7H6N2O Formula Weight: 134.14 Melting Point: 314-316° Beilstein Reference: 119866

Formula: C₇H₆N₂O
Melting Pt: 314…316 °C
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00127894
CAS Number: 615-16-7
EINECS: 210-412-4
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR