Sudan III
Supplier: Thermo Scientific Chemicals
A18318-14
A18318-22
A18318-36
CAAAA18318-14EA
48.66
CAD
CAAAA18318-14
CAAAA18318-22
CAAAA18318-36
Sudan III
Sudan III
CAS No.: 85-86-9
Molecular Formula: C22H16N4O
Formula Weight: 352.40
Melting Point: ca 200° dec.
Beilstein Reference: 931185
Merck Reference: 14,8884
MDL No.: MFCD00003905
Formula:
C₂₂H₁₆N₄O Melting Pt: ca 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003905 CAS Number: 85-86-9 EINECS: 201-638-4 Merck Index: 14,8884 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...