5-Nitro-α,α,α-trifluoro-m-toluic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Nitro-5-(trifluoromethyl)benzoic acid
A18316-06
A18316-14
CAAAA18316-06EA
66.99
CAD
CAAAA18316-06
CAAAA18316-14
5-Nitro-α,α,α-trifluoro-m-toluic acid 97%
5-Nitro-α,α,α-trifluoro-m-toluic acid
Purity: 97%
CAS No.: 328-80-3
Molecular Formula: C8H4F3NO4
Formula Weight: 235.12
Melting Point: 127-132°
Beilstein Reference: 2284651
Formula:
C₈H₄F₃NO₄ MW: 235.12 g/mol Melting Pt: 127…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024509 CAS Number: 328-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...