D-(+)-Raffinose pentahydrate 99%
Supplier: Thermo Scientific Chemicals
A18313-09
A18313-18
CAAAA18313-09EA
67.15
CAD
CAAAA18313-09
CAAAA18313-18
D-(+)-Raffinose pentahydrate 99%
D(+)-Raffinose pentahydrate
Purity: 99%
CAS No.: 17629-30-0
Molecular Formula: C18H32O16.5H2O
Formula Weight: 594.52 (504.45anhy)
Physical Form: Cryst powder
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 78-82°
Beilstein Reference: 4285763
Merck Reference: 14,8096
Optical Rotation: +105° (c=10 in water)
MDL No.: MFCD00071590
Formula:
C₁₈H₃₂O₁₆·5H₂O Melting Pt: 78…82 °C Density: 1.516 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071590 CAS Number: 17629-30-0 EINECS: 208-146-9 Merck Index: 14,8096 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...